3-Amino-3-(2,3-dichlorophenyl)propionic acid

Leverantör: Apollo Scientific

Synonyms: 3-Amino-3-(2,3-dichlorophenyl)propanoic acid

OR14360-1G OR14360-5G
APOSOR14360-1GEA 1725 SEK
APOSOR14360-1G APOSOR14360-5G
3-Amino-3-(2,3-dichlorophenyl)propionic acid
3-Amino-3-(2,3-dichlorophenyl)propionic acid
Formel: C₉H₉Cl₂NO₂
MW: 234,0800 g/mol
Smältpunkt: 232…235 °C
Storage Temperature: Omgivande
MDL Number: MFCD01871301
CAS nummer: 117391-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR