4-Methyloxazole-5-carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
OR14298-10G OR14298-25G OR14298-5G
APOSOR14298-10GEA 1800 SEK
APOSOR14298-10G APOSOR14298-25G APOSOR14298-5G
4-Methyloxazole-5-carboxylic acid 97%
4-Methyloxazole-5-carboxylic acid
Formel: C₅H₅NO₃
MW: 127,1000 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01571070
CAS nummer: 2510-32-9