1-Boc-4-(2-aminophenyl)piperazine

Leverantör: Apollo Scientific
Varning

Synonyms: 4-(2-Aminophenyl)piperazine, N1-BOC protected, 1-Boc-4-(2-Aminophenyl)piperazine

OR14260-1G OR14260-250MG
APOSOR14260-1GEA 1050 SEK
APOSOR14260-1G APOSOR14260-250MG
1-Boc-4-(2-aminophenyl)piperazine
1-Boc-4-(2-aminophenyl)piperazine
Formel: C₁₅H₂₃N₃O₂
MW: 277,368 g/mol
Smältpunkt: 120…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD04115046
CAS nummer: 170017-74-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR