6-Bromoimidazo[1,2-a]pyridine

Leverantör: Apollo Scientific
Varning

OR1419-5G OR1419-25G
APOSOR1419-5GEA 210 SEK
APOSOR1419-5G APOSOR1419-25G
6-Bromoimidazo[1,2-a]pyridine
6-Bromoimidazo[1,2-a]pyridine
Formel: C₇H₅BrN₂
MW: 197,03 g/mol
Smältpunkt: 75…80 °C
Storage Temperature: Omgivande
MDL Number: MFCD06496200
CAS nummer: 6188-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR