N-Benzyl-2-cyanoacetamide

Leverantör: Apollo Scientific
Varning

OR14083-1G OR14083-5G
APOSOR14083-1GEA 225 SEK
APOSOR14083-1G APOSOR14083-5G
N-Benzyl-2-cyanoacetamide
N-Benzyl-2-cyanoacetamide
Formel: C₁₀H₁₀N₂O
Smältpunkt: 120…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00461336
CAS nummer: 10412-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR