5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine

Leverantör: Apollo Scientific
Varning

Synonyms: 5-(4-Pyridyl)-1,3,4-thiadiazol-2-amine, 2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole

OR13740-100G OR13740-25G
APOSOR13740-100GEA 12400 SEK
APOSOR13740-100G APOSOR13740-25G
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine
5-Pyridin-4-yl[1,3,4]thiadiazol-2-ylamine
Formel: C₇H₆N₄S
MW: 178,22 g/mol
Smältpunkt: 242…248 °C
Storage Temperature: Omgivande
MDL Number: MFCD00473692
CAS nummer: 2002-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR