5-Bromo-1,3-dihydrobenzoimidazol-2-one
Leverantör: Apollo Scientific
Varning
OR13660-1G OR13660-5G
APOSOR13660-1GEA 875 SEK
APOSOR13660-1G APOSOR13660-5G
5-Bromo-1,3-dihydrobenzoimidazol-2-one
5-Bromo-1,3-dihydrobenzoimidazol-2-one
Formel: C₇H₅BrN₂O
MW: 213,04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01893032
CAS nummer: 39513-26-3