Atropic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Phenylacrylic Acid, Ipratropium bromide Impurity D
OR13658-25G OR13658-1G OR13658-5G
APOSOR13658-25GEA 1075 SEK
APOSOR13658-25G APOSOR13658-1G APOSOR13658-5G
Atropic acid 98%
Atropic acid
Formel: C₉H₈O₂
MW: 148,16 g/mol
Kokpunkt: 267 °C
Smältpunkt: 107 °C
Storage Temperature: Omgivande
MDL Number: MFCD00046531
CAS nummer: 492-38-6
EINECS: 207-753-6