2-Amino-4,6-dichloropyrimidine-5-carbaldehyde
Leverantör: Apollo Scientific
|
Varning
|
OR13645-1G
OR13645-10G
OR13645-25G
APOSOR13645-1GEA
225
SEK
APOSOR13645-1G
APOSOR13645-10G
APOSOR13645-25G
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde
Formel:
C₅H₃Cl₂N₃O MW: 192,00 g/mol Smältpunkt: 208…224 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD03001242 CAS nummer: 5604-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...