4-Ethyl-2,3-dioxo-1-piperazinecarbonyl chloride
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
4-Ethyl-2,3-dioxopiperazine-1-carbonyl chloride, 2,3-Dioxo-4-ethylpiperazine-1-carbonyl chloride
OR13641-5G
OR13641-25G
OR13641-100G
APOSOR13641-5GEA
1200
SEK
APOSOR13641-5G
APOSOR13641-25G
APOSOR13641-100G
4-Ethyl-2,3-dioxo-1-piperazinecarbonyl chloride
4-Ethyl-2,3-dioxo-1-piperazinecarbonyl chloride
Formel:
C₇H₉ClN₂O₃ MW: 204,61 g/mol Smältpunkt: 95…96 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00067054 CAS nummer: 59703-00-3 EINECS: 261-867-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...