Glykolsyra 98%

Leverantör: Apollo Scientific
Fara

OR13191-100G OR13191-500G
APOSOR13191-100GEA 890 SEK
APOSOR13191-100G APOSOR13191-500G
Glykolsyra 98%
Glykolsyra
Formel: C₂H₄O₃
Smältpunkt: 74…80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004312
CAS nummer: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR