N-Benzylbenzamide 98%
Leverantör: Apollo Scientific
|
Varning
|
OR1310-25G
OR1310-100G
APOSOR1310-25GEA
1725
SEK
APOSOR1310-25G
APOSOR1310-100G
N-Benzylbenzamide 98%
N-Benzylbenzamide
Formel:
C₁₄H₁₃NO MW: 211,2591 g/mol Smältpunkt: 104…108 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00003070 CAS nummer: 1485-70-7 EINECS: 216-063-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...