4-Iodo-3-nitrophenol

Leverantör: Apollo Scientific
Varning

OR13076-25G OR13076-5G
APOSOR13076-25GEA 7225 SEK
APOSOR13076-25G APOSOR13076-5G
4-Iodo-3-nitrophenol
4-Iodo-3-nitrophenol
Formel: C₆H₄INO₃
MW: 265,01 g/mol
Smältpunkt: 156 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320680
CAS nummer: 113305-56-9
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR