5-Bromopicolinic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid
OR13059-25G OR13059-5G
APOSOR13059-25GEA 585 SEK
APOSOR13059-25G APOSOR13059-5G
5-Bromopicolinic acid 97%
5-Bromopicolinic acid
Formel: C₆H₄BrNO₂
MW: 202.01 g/mol
Smältpunkt: 173…175 °C
Storage Temperature: Omgivande
MDL Number: MFCD00234149
CAS nummer: 30766-11-1