(3,4-Dichlorophenyl)methanesulphonyl chloride, Technical Grade
Leverantör: Apollo Scientific
Fara
OR12683-1G OR12683-250MG
APOSOR12683-1GEA 325 SEK
APOSOR12683-1G APOSOR12683-250MG
(3,4-Dichlorophenyl)methanesulphonyl chloride, Technical Grade
(3,4-Dichlorophenyl)methanesulphonyl chloride
Formel: C₇H₅CI₃O₂S
MW: 259,54 g/mol
Smältpunkt: 65…69 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631929
CAS nummer: 85952-30-3
UN: 3261
ADR: 8,II