3-Benzylpyrrolidine

Leverantör: Apollo Scientific
Varning

OR12550-1G OR12550-250MG
APOSOR12550-1GEA 7375 SEK
APOSOR12550-1G APOSOR12550-250MG
3-Benzylpyrrolidine
3-Benzylpyrrolidine
Formel: C₁₁H₁₅N
Storage Temperature: Kylskåp
MDL Number: MFCD01862537
CAS nummer: 170304-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR