4-Chloro-1-benzothiophene-2-carbohydrazide, Technical Grade
Leverantör: Apollo Scientific
Varning
OR12457-1G OR12457-5G OR12457-250MG
APOSOR12457-1GEA 550 SEK
APOSOR12457-1G APOSOR12457-5G APOSOR12457-250MG
4-Chloro-1-benzothiophene-2-carbohydrazide, Technical Grade
4-Chloro-1-benzothiophene-2-carbohydrazide
Formel: C₉H₇ClN₂OS
MW: 226,69 g/mol
Smältpunkt: 184…186 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09817458
CAS nummer: 952182-46-6