DL-α-(4-Hydroxyphenyl)glycine
Leverantör: Apollo Scientific
Varning
Synonyms: H-DL-2-Phg(4-OH)-OH, 2-Amino-2-(4-hydroxyphenyl)acetic acid
OR12450-1G OR12450-250MG OR12450-2.5G OR12450-5G
APOSOR12450-1GEA 685 SEK
APOSOR12450-1G APOSOR12450-250MG APOSOR12450-2.5G APOSOR12450-5G
DL-α-(4-Hydroxyphenyl)glycine
DL-α-(4-Hydroxyphenyl)glycine
Formel: C₈H₉NO₃
MW: 167,1600 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00065931
CAS nummer: 938-97-6