4-Ethylcyclohexylamine (cis- and trans- mixture) 95%
Leverantör: Apollo Scientific
Fara
Synonyms: 1-Amino-4-ethylcyclohexane
OR12347-10G OR12347-5G
APOSOR12347-10GEA 6250 SEK
APOSOR12347-10G APOSOR12347-5G
4-Ethylcyclohexylamine (cis- and trans- mixture) 95%
4-Ethylcyclohexylamine (cis- and trans- mixture)
Formel: C₈H₁₇N
MW: 127,23 g/mol
Kokpunkt: 75…80 °C/20mmHg
Densitet: 0,86 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD04969087
CAS nummer: 42195-97-1
UN: 2924
ADR: 3+8,III