Pyrazin-2-ylmethanol, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Pyrazinemethanol, 2-(Hydroxymethyl)pyrazine
OR12341-1G OR12341-5G OR12341-10G
APOSOR12341-1GEA 310 SEK
APOSOR12341-1G APOSOR12341-5G APOSOR12341-10G
Pyrazin-2-ylmethanol, Technical Grade
Pyrazin-2-ylmethanol
Formel: C5H6N2O
MW: 110,11 g/mol
Smältpunkt: 32…36 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01693938
CAS nummer: 6705-33-5