1,3-benzothiazol-5-amine 95%

Leverantör: Apollo Scientific
Varning

Synonyms: 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole

OR12328-1G
APOSOR12328-1GEA 260 SEK
APOSOR12328-1G
1,3-benzothiazol-5-amine 95%
1,3-benzothiazol-5-amine
Formel: C₇H₆N₂S
MW: 150.199 g/mol
Smältpunkt: 73…78 °C
Storage Temperature: Kylskåp
MDL Number: MFCD04115282
CAS nummer: 1123-93-9
EINECS: 214-381-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR