2-(Morpholinosulphonyl)phenylboronic acid
Leverantör: Apollo Scientific
|
Varning
|
OR11971-1G
OR11971-5G
OR11971-100MG
APOSOR11971-1GEA
2750
SEK
APOSOR11971-1G
APOSOR11971-5G
APOSOR11971-100MG
2-(Morpholinosulphonyl)phenylboronic acid
2-(Morpholinosulphonyl)phenylboronic acid
Formel:
C₁₀H₁₄BNO₅S Storage Temperature: Omgivande |
MDL Number:
MFCD09878327 CAS nummer: 957062-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...