Xanthone

Leverantör: Apollo Scientific
Varning

Synonyms: Xanthen-9-one, 9H-Xanthen-9-one

OR1180-100G OR1180-500G
APOSOR1180-100GEA 1200 SEK
APOSOR1180-100G APOSOR1180-500G
Xanthone
Xanthone
Formel: C₁₃H₈O₂
MW: 196,21 g/mol
Kokpunkt: 349…350 °C/730mmHg
Smältpunkt: 174…176 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005060
CAS nummer: 90-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR