4-Hydroxybenzyl alcohol 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol
OR1172-250G OR1172-50G
APOSOR1172-250GEA 650 SEK
APOSOR1172-250G APOSOR1172-50G
4-Hydroxybenzyl alcohol 98%
4-Hydroxybenzyl alcohol
Formel: C₇H₈O₂
MW: 124,14 g/mol
Kokpunkt: 251…253 °C
Smältpunkt: 110…112 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00004658
CAS nummer: 623-05-2
EINECS: 210-768-0