Methyl-6-amino-3-bromopicolinate

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 6-amino-3-bromopicolinate, Methyl 6-amino-3-bromopyridine-2-carboxylate

OR11647-1G OR11647-5G
APOSOR11647-1GEA 910 SEK
APOSOR11647-1G APOSOR11647-5G
Methyl-6-amino-3-bromopicolinate
Methyl-6-amino-3-bromopicolinate
Formel: C₇H₇BrN₂O₂
MW: 231,0467 g/mol
Smältpunkt: 95…97 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09800897
CAS nummer: 178876-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR