4-Bromo-N,N-3-trimethylbenzenesulphonamide 98%
Leverantör: Apollo Scientific
Varning
OR11443-1G OR11443-5G
APOSOR11443-1GEA 1775 SEK
APOSOR11443-1G APOSOR11443-5G
4-Bromo-N,N-3-trimethylbenzenesulphonamide 98%
4-Bromo-N,N-3-trimethylbenzenesulphonamide
Formel: C₉H₁₂BrNO₂S
Storage Temperature: Omgivande
MDL Number: MFCD09475820
CAS nummer: 849532-31-6