3-Methyl-2-nitroanisole 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Methoxy-2-nitrotoluene, 1-Methoxy-3-methyl-2-nitrobenzene, Methyl 2-nitro-m-tolyl ether

OR11213-10G OR11213-25G OR11213-5G
APOSOR11213-10GEA 190 SEK
APOSOR11213-10G APOSOR11213-25G APOSOR11213-5G
3-Methyl-2-nitroanisole 97%
3-Methyl-2-nitroanisole
Formel: C₈H₉NO₃
MW: 167,16 g/mol
Kokpunkt: 170 °C/35mmHg
Smältpunkt: 48…52 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007179
CAS nummer: 5345-42-6
EINECS: 226-295-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR