1-tert-Butyl-3-methyl-4-oxopiperidine-1,3-dicarboxylate
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1-tert-Butyl 3-methyl 4-oxopiperidine-1,3-dicarboxylate
OR111577-1G
OR111577-5G
APOSOR111577-1GEA
345
SEK
APOSOR111577-1G
APOSOR111577-5G
1-tert-Butyl-3-methyl-4-oxopiperidine-1,3-dicarboxylate
1-tert-Butyl-3-methyl-4-oxopiperidine-1,3-dicarboxylate
Formel:
C₁₂H₁₉NO₅ MW: 257,2830 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD08752608 CAS nummer: 161491-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...