Ethyl-6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate

Leverantör: Apollo Scientific
Varning

OR111531-1G OR111531-5G
APOSOR111531-1GEA 175 SEK
APOSOR111531-1G APOSOR111531-5G
Ethyl-6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₁H₁₁IN₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD09152737
CAS nummer: 952183-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR