Etynylcyklopropan 98%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
Cyclopropylacetylene
OR11148-100G
OR11148-25G
OR11148-5G
APOSOR11148-100GEA
2225
SEK
APOSOR11148-100G
APOSOR11148-25G
APOSOR11148-5G
Etynylcyklopropan 98%
Etynylcyklopropan
Formel:
C₅H₆ MW: 66,1 g/mol Kokpunkt: 51 °C Densitet: 0,781 g/cm³ (25 °C) Flampunkt: –17 °C Storage Temperature: Frys |
MDL Number:
MFCD02181090 CAS nummer: 6746-94-7 EINECS: 425-430-1 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...