6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR111308-250MG OR111308-500MG
APOSOR111308-250MGEA 175 SEK
APOSOR111308-250MG APOSOR111308-500MG
6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid
6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid
Formel: C₈H₆ClN₃O₂
MW: 211,6051 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08704709
CAS nummer: 14714-22-8