1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%
Leverantör: Apollo Scientific
Varning
Synonyms: 1,1-Bis(chloromethyl)-2,2-dibromocyclopropane
OR11129-10G OR11129-5G
APOSOR11129-10GEA 740 SEK
APOSOR11129-10G APOSOR11129-5G
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane
Formel: C₅H₆Br₂Cl₂
MW: 296,81 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00101445
CAS nummer: 98577-44-7