Thieno[3,2-b]pyridine-2-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR111149-1G OR111149-250MG
APOSOR111149-1GEA 2125 SEK
APOSOR111149-1G APOSOR111149-250MG
Thieno[3,2-b]pyridine-2-carboxylic acid
Thieno[3,2-b]pyridine-2-carboxylic acid
Formel: C₈H₅NO₂S
MW: 179.2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12198115
CAS nummer: 347187-30-8