(±)-1-Aminoindan

Leverantör: Apollo Scientific
Varning

Synonyms: 1-Aminoindan, 1-Indanamine, 2,3-Dihydro-1H-inden-1-amine, (±)-1-Indanylamine

OR11114-1G OR11114-5G
APOSOR11114-1GEA 395 SEK
APOSOR11114-1G APOSOR11114-5G
(±)-1-Aminoindan
(±)-1-Aminoindan
Formel: C₉H₁₁N
MW: 133.19 g/mol
Kokpunkt: 96…98 °C
Smältpunkt: 1.5 °C
Densitet: 1.038 g/cm³ (25 °C)
Flampunkt: 94 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003799
CAS nummer: 34698-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR