3-Chloroanthranilic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Amino-3-chlorobenzoic acid
OR11113-100G OR11113-25G
APOSOR11113-100GEA 755 SEK
APOSOR11113-100G APOSOR11113-25G
3-Chloroanthranilic acid 98%
3-Chloroanthranilic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Smältpunkt: 189…195 °C
Storage Temperature: Omgivande
MDL Number: MFCD00134229
CAS nummer: 6388-47-2