4-(1-Pyrazolyl)benzonitrile

Leverantör: Apollo Scientific
Varning

Synonyms: 1-(4-Cyanophenyl)pyrazole

OR111078-10G OR111078-1G OR111078-5G
APOSOR111078-10GEA 1400 SEK
APOSOR111078-10G APOSOR111078-1G APOSOR111078-5G
4-(1-Pyrazolyl)benzonitrile
4-(1H-Pyrazol-3-yl)benzonitrile
Formel: C₁₀H₇N₃
MW: 169,19 g/mol
Smältpunkt: 140…142 °C
Storage Temperature: Omgivande
MDL Number: MFCD09033533
CAS nummer: 25699-83-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR