1,3-Benzodioxole-5,6-diamine, Technical Grade
Leverantör: Apollo Scientific
Varning
OR110848-100MG OR110848-1G OR110848-500MG
APOSOR110848-100MGEA 1150 SEK
APOSOR110848-100MG APOSOR110848-1G APOSOR110848-500MG
1,3-Benzodioxole-5,6-diamine, Technical Grade
1,3-Benzodioxole-5,6-diamine
Formel: C₇H₈N₂O₂
Smältpunkt: 95…97 °C (dec.)
Storage Temperature: Frys
MDL Number: MFCD07780137
CAS nummer: 38608-07-0