2',5'-Dichloroacetophenone

Leverantör: Apollo Scientific
Varning

OR11050-100G OR11050-25G
APOSOR11050-100GEA 790 SEK
APOSOR11050-100G APOSOR11050-25G
2',5'-Dichloroacetophenone
2',5'-Dichloroacetophenone
Formel: C₈H₆Cl₂O
MW: 189,04 g/mol
Kokpunkt: 118 °C/12mmHg
Smältpunkt: 41579 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000607
CAS nummer: 2476-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR