Ethyl-3-Amino-2-methylbenzoate
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Amino-o-toluic Acid Ethyl Ester, 3-Amino-2-methylbenzoic Acid Ethyl Ester, Ethyl 3-amino-2-methylbenzoate, Ethyl 3-Amino-o-toluate
OR110425-1G OR110425-25G OR110425-5G
APOSOR110425-1GEA 210 SEK
APOSOR110425-1G APOSOR110425-25G APOSOR110425-5G
Ethyl-3-Amino-2-methylbenzoate
Ethyl-3-Amino-2-methylbenzoate
Formel: C₁₀H₁₃NO₂
MW: 179,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00957246
CAS nummer: 57414-85-4