2-Amino-5-chloro-3-iodopyridine

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Chloro-3-iodo-2-pyridinamine, 5-Chloro-3-iodo-2-pyridylamine, 5-Chloro-3-iodopyridin-2-amine

OR110378-5G OR110378-10G OR110378-25G
APOSOR110378-5GEA 1150 SEK
APOSOR110378-5G APOSOR110378-10G APOSOR110378-25G
2-Amino-5-chloro-3-iodopyridine
2-Amino-5-chloro-3-iodopyridine
Formel: C₅H₄ClIN₂
MW: 254,46 g/mol
Smältpunkt: 85…87 °C
Storage Temperature: Omgivande
MDL Number: MFCD07644578
CAS nummer: 211308-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR