Pyrazine-2-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid
OR11036-100G OR11036-250G OR11036-500G
APOSOR11036-100GEA 550 SEK
APOSOR11036-100G APOSOR11036-250G APOSOR11036-500G
Pyrazine-2-carboxylic acid
Pyrazine-2-carboxylic acid
Formel: C₅H₄N₂O₂
MW: 124,1 g/mol
Smältpunkt: 222…225 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006130
CAS nummer: 98-97-5
EINECS: 202-718-1