1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Leverantör: Apollo Scientific
Varning
OR110357-10G OR110357-1G OR110357-5G
APOSOR110357-10GEA 6725 SEK
APOSOR110357-10G APOSOR110357-1G APOSOR110357-5G
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
1,4,6-Trimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formel: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00829059
CAS nummer: 64038-03-5