(2E)-1-(3,4-Dimethoxyphenyl)-3-(dimethylamino)prop-2-en-1-one
Leverantör: Apollo Scientific
OR110249-1G OR110249-10G OR110249-5G
APOSOR110249-1GEA 1100 SEK
APOSOR110249-1G APOSOR110249-10G APOSOR110249-5G
(2E)-1-(3,4-Dimethoxyphenyl)-3-(dimethylamino)prop-2-en-1-one
(2E)-1-(3,4-Dimethoxyphenyl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₃H₁₇NO₃
MW: 235,29 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02187201
CAS nummer: 127172-22-9