1H-Pyrazole-4-boronic acid pinacol ester
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR11023-5G OR11023-25G OR11023-100G
APOSOR11023-5GEA 260 SEK
APOSOR11023-5G APOSOR11023-25G APOSOR11023-100G
1H-Pyrazole-4-boronic acid pinacol ester
1H-Pyrazole-4-boronic acid pinacol ester
Formel: C₉H₁₅BN₂O₂
MW: 194.039 g/mol
Smältpunkt: 142…149 °C
Storage Temperature: Omgivande
MDL Number: MFCD03453063
CAS nummer: 269410-08-4