2-Butyl-5,6-dichloro-1H-1,3-benzimidazole
Leverantör: Apollo Scientific
OR110228-1G OR110228-5G
APOSOR110228-1GEA 1275 SEK
APOSOR110228-1G APOSOR110228-5G
2-Butyl-5,6-dichloro-1H-1,3-benzimidazole
2-Butyl-5,6-dichloro-1H-1,3-benzimidazole
Formel: C₁₁H₁₂Cl₂N₂
Storage Temperature: Omgivande
MDL Number: MFCD08064759
CAS nummer: 133052-59-2