Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate
Leverantör: Apollo Scientific
OR110188-1G OR110188-5G
APOSOR110188-1GEA 1800 SEK
APOSOR110188-1G APOSOR110188-5G
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate
Methyl-3-[(3-chloro-2-formylphenyl)thio]propanoate
Formel: C₁₁H₁₁ClO₃S
MW: 258,73 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11100102
CAS nummer: 1215582-82-3