1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22)
Leverantör: Apollo Scientific
Varning
Synonyms: Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
OR10939-1G OR10939-5G
APOSOR10939-1GEA 1725 SEK
APOSOR10939-1G APOSOR10939-5G
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22)
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22)
Formel: C₁₂H₂₆N₂O₄
MW: 262,35 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005112
CAS nummer: 23978-55-4