Tetraethyl methylenebisphosphonate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)
OR10799-1G OR10799-25G OR10799-5G
APOSOR10799-1GEA 210 SEK
APOSOR10799-1G APOSOR10799-25G APOSOR10799-5G
Tetraethyl methylenebisphosphonate 97%
Tetraethyl methylenebisphosphonate
Formel: C₉H₂₂O₆P₂
MW: 288,22 g/mol
Kokpunkt: 171…174 °C
Densitet: 1,162 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039887
CAS nummer: 1660-94-2
EINECS: 216-764-5