2,6-Dibromo-4-tert-butylaniline 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(tert-Butyl)-2,6-dibromoaniline, 2,6-Dibromo-4-(tert-butyl)aniline
OR1068-100G OR1068-25G OR1068-5G
APOSOR1068-100GEA 3175 SEK
APOSOR1068-100G APOSOR1068-25G APOSOR1068-5G
2,6-Dibromo-4-tert-butylaniline 97%
2,6-Dibromo-4-tert-butylaniline
Formel: C₁₀H₁₃Br₂N
MW: 307,03 g/mol
Kokpunkt: 136…140 °C/2mmHg
Storage Temperature: Kylskåp
MDL Number: MFCD06657952
CAS nummer: 10546-67-5