(Triphenylphosphoranylidene)acetonitrile
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(Triphenylphosphoranylidene)acetonitrile, (Cyanomethylene)triphenylphosphorane
OR10639-25G OR10639-5G OR10639-100G
APOSOR10639-25GEA 855 SEK
APOSOR10639-25G APOSOR10639-5G APOSOR10639-100G
(Triphenylphosphoranylidene)acetonitrile
(Triphenylphosphoranylidene)acetonitrile
Formel: C₂₀H₁₆NP
MW: 301,33 g/mol
Smältpunkt: 189…198 °C
Storage Temperature: Omgivande
MDL Number: MFCD00567633
CAS nummer: 16640-68-9
EINECS: 240-689-7